Double spatulas, micro
130 mm, 4 mm
Katalog # 231-0427
Leverandør: RSG
Gå til:
Product Details & Documents
Stainless steel. Anti-magnetic, very flexible micro powder spatula.
- With round handle, one flat and one scoop shaped, rounded spatula end
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Længde:130 mm
- Bladbredde:4 mm