Orders LinkContactUsLinkComponent
Accessories for heating circulators
Accessories for heating circulators
2× adapters, M16×1 female to NPT ⅜" male
Katalog # JULA8890006
Leverandør:   JULABO GmbH
Accessories for heating circulators
Katalog # JULA8890006
Leverandør:  JULABO GmbH
Accessories for heating circulators
Accessories for heating circulators
2× adapters, M16×1 female to NPT ⅜" male
Price
748,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    2× adapters, M16×1 female to NPT ⅜" male