Test sets
Test set for conductive compounds
Katalog # DRAE8103170
Leverandør: Dräger
Gå til:
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifikationer
- Beskrivelse:Test set for conductive compounds
- Seq:04
- Cat. No.:DRAE8103170
- Model:Test set for conductive compounds
- Måletid:120 s