Orders LinkContactUsLinkComponent
m-Endo broth
m-Endo broth, 2 ml plastic ampule, E. coli and coliforms, 50
Katalog # HACH2373550
Leverandør:   Hach
m-Endo broth
Katalog # HACH2373550
Leverandør:  Hach
CAS nummer:  
m-Endo broth
m-Endo broth
m-Endo broth, 2 ml plastic ampule, E. coli and coliforms, 50
CAS nummer:  
Price
987,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
m-Endo broth detects total coliform bacteria (colonies with a greenish-gold sheen). For potable water samples, confirm with Lauryl Tryptose and brilliant green bile broth tubes. Samples that test positive for total coliform bacteria should be further analysed for the presence of faecal coliform or E. coli using EC Medium or EC Medium with MUG respectively.
  • Save valuable time and money using prepared media for membrane filtration methods
Specifikationer
Specifikationer
  • Pakn:
    50
  • Beskrivelse:
    m-Endo broth, 2 ml plastic ampule
  • Til:
    E. coli and coliforms