TLC polyester sheets
POLYGRAM® TLC polyester sheets. Kit includes 100 × SIL G/UV254, 50 × Alox N/UV254, 50 × CEL 300, 40×80 mm, F254
Katalog # 552-0280
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
Documents
Thin Layer Chromatography.pdf
Yderligere produktdetaljer
Specifikationer
- Pladestørrelse:40×80 mm
- Beskrivelse:POLYGRAM® TLC polyester sheets. Kit includes 100 × SIL G/UV254, 50 × Alox N/UV254, 50 × CEL 300
- Fluorescensindikator:F254
- Cat. No.:552-0280