Orders LinkContactUsLinkComponent
TLC introductory kit, Micro Set A
TLC introductory kit, Micro Set A
Collection of 7 indiviudual components of test dye mixture 2
Katalog # MANA814012
Leverandør:   MACHEREY-NAGEL
TLC introductory kit, Micro Set A
Katalog # MANA814012
Leverandør:  MACHEREY-NAGEL
TLC introductory kit, Micro Set A
TLC introductory kit, Micro Set A
Collection of 7 indiviudual components of test dye mixture 2
Price
902,00 kr
Log ind for tilgængelighed
1 * 1 Each
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
An ideal introductory kit for the science education sector. The kit contains all the the chemicals, plates and accessories for students to perform separations of dyes including: anthaquinone dye mixtures, lipophilic dye mixtures and food dye mixtures. The TLC plates are polyester sheet based with either silica, cellulose or alumina as coating.
Leveringsinformation: Kit includes 3 developing tanks, 50 glass capillaries (1 µl), 1 spotting guide, 1×10 ml measuring cylinder, 50 sheets 40×80 mm POLYGRAM SIL G/UV₂₅₄, 50 sheets 40×80 mm ALOX N/UV₂₅₄ and 50 sheets 40×80 mm of CEL 300, 3 dye mixtures (8 ml), 3 prepared solvent mixtures (100 ml), 2 felt pens and instruction manual.
Specifikationer
  • Beskrivelse:
    Collection of 7 indiviudual components of test dye mixture 2
  • Seq:
    03