Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Qualitative Test Papers, Cobalt

Cobalt test paper for cobalt, 25 - mg/L - Co²⁺, Complexation with rhodanide ions, 48×25×84 mm (packaging), 100 Tests, 100
Katalog # MANA90728
Leverandør:   MACHEREY-NAGEL
Qualitative Test Papers, Cobalt
Katalog # MANA90728
Leverandør:  MACHEREY-NAGEL
CAS nummer:  
Qualitative Test Papers, Cobalt
Qualitative Test Papers, Cobalt

Cobalt test paper for cobalt, 25 - mg/L - Co²⁺, Complexation with rhodanide ions, 48×25×84 mm (packaging), 100 Tests, 100

CAS nummer:  
Price
335,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips are ideal to test levels of phosphate in solutions. The easy 'dip and read' procedure provides realiable results within 90 seconds.

Specifikationer

Specifikationer

  • Kemisk metode:
    Complexation with rhodanide ions
  • Pakn:
    100
  • Beskrivelse:
    Cobalt test paper for cobalt
  • Dimensioner:
    48×25×84 mm (packaging)
  • Test pr. sæt:
    100 Tests
  • Område:
    25 - mg/L - Co²⁺
  • Indhold:
    100 test strips in a plastic box
  • Cat. No.:
    MANA90728
  • Sensitivitet:
    Cobalt - 25 mg/L Co²⁺
  • Overensstemmelse:
    Not CE certified and no hazardous material
  • Platform:
    Qualitative test paper
  • B×D×H:
    20×70 mm (net)
  • Holdbarhed (måneder):
    24 (from production)
  • Vægt:
    46,5 g
  • Method:
    Cobalt - white → blue

Specifikationer