VISOCOLOR® Colorimetric Test, Phosphate
VISOCOLOR® alpha Phosphate, Reagents for 70 tests and color chart in a blister, 2 • 5 • 10 • 15 • 20 mg/L (PO₄³⁻), Phosphomolybdenum blue, Visual, 154×93×35 mm (Including packaging), 70, 1 KIT
Reagents for the determination of phosphate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
- Kemisk metode:Phosphomolybdenum blue
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® alpha Phosphate
- Dimensioner:154×93×35 mm (Including packaging)
- Til enhed:Visual
- Test pr. sæt:70
- Område:2 • 5 • 10 • 15 • 20 mg/L (PO₄³⁻)
- Indhold:Reagents for 70 tests and color chart in a blister
- Cat. No.:MANA935079
- Platform:Colorimetric test kits
- Holdbarhed (måneder):24
- Vægt:99,6 g (Including packaging)
- Opbevaringstemperatur:15...25 °C



