Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, pH

VISOCOLOR® Colorimetric Test, pH

VISOCOLOR® alpha pH 5 - 9, Reagents for 200 tests and color chart in a blister, 5.0 • 5.5 • 6.0 • 6.5 • 7.0 • 7.5 • 8.0 • 8.5 • 9.0 pH, Mixed indicator, Visual, 154×93×35 mm (including packaging), Hazardous material: Yes, 200, 1 KIT
Catalog # MANA935075
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, pH
Catalog # MANA935075
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, pH
VISOCOLOR® Colorimetric Test, pH

VISOCOLOR® alpha pH 5 - 9, Reagents for 200 tests and color chart in a blister, 5.0 • 5.5 • 6.0 • 6.5 • 7.0 • 7.5 • 8.0 • 8.5 • 9.0 pH, Mixed indicator, Visual, 154×93×35 mm (including packaging), Hazardous material: Yes, 200, 1 KIT

CAS Number:  
Price
338,00 kr
Log in for availability
1 * 1 Each
Specifications

Specifications

  • Kemisk metode:
    Mixed indicator
  • Pakn:
    1 KIT
  • Beskrivelse:
    VISOCOLOR® alpha pH 5 - 9
  • Dimensioner:
    154×93×35 mm (including packaging)
  • Model:
    Hazardous material: Yes
  • Til enhed:
    Visual
  • Test pr. sæt:
    200
  • Område:
    5.0 • 5.5 • 6.0 • 6.5 • 7.0 • 7.5 • 8.0 • 8.5 • 9.0 pH
  • Indhold:
    Reagents for 200 tests and color chart in a blister
  • Cat. No.:
    MANA935075
  • Platform:
    Colorimetric test kits
  • Holdbarhed (måneder):
    36
  • Vægt:
    53,3 g (including packaging)
  • Model:
    MANA935075
  • Opbevaringstemperatur:
    15…25 °C

Specifications