VISOCOLOR® Colorimetric Test, Ammonia
VISOCOLOR® alpha Ammonium, Reagents for 50 tests and colour chart in a blister, 0 • 0,2 • 0,5 • 1 • 2 • 3 mg/L (NH₄⁺), Indophenol, Visual, 190×92×34 mm (Including packaging), Sea water compatible, 50, 1 KIT
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
Reagents for the determination of ammonia in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Hazardous material: Yes
Documents
Certifikater
Søg efter partinummerIFU VISOCOLOR® Alpha Ammonium NH₄⁺.pdf IFU VISOCOLOR® ECO Ammonium 15.pdf IFU VISOCOLOR® ECO Ammonium 3.pdf IFU VISOCOLOR® HE Ammonium.pdf IFU_Ammonium 15 ECO Visocolor
More
Specifikationer
Specifikationer
- Kemisk metode:Indophenol
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® alpha Ammonium
- Dimensioner:190×92×34 mm (Including packaging)
- Model:Sea water compatible
- Til enhed:Visual
- Test pr. sæt:50
- Område:0 • 0,2 • 0,5 • 1 • 2 • 3 mg/L (NH₄⁺)
- Indhold:Reagents for 50 tests and colour chart in a blister
- Cat. No.:MANA935012
- Platform:Colorimetric test kits
- Holdbarhed (måneder):18
- Vægt:110,5 g (Including packaging)
- Model:MANA935012
- Opbevaringstemperatur:15...25 °C
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



