VISOCOLOR® Colorimetric Test, Phosphate
VISOCOLOR® ECO Phosphate, refill pack, test no. 5-84; Measuring range on PF-12Plus. Range on other photometers can be different., Reagents for 80 tests in a box, 0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 • 5 mg/L (PO₄ - P); 0,2 - 5,0 mg/L (PO₄ - P), Phosphomolybdenum blue, PF-12Plus;PF-3 Soil;Advance;PF-3 Fish;Visual, 170×115×70 mm (Including packaging), 80, 1 KIT
Reagents for the determination of phosphate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
- Kemisk metode:Phosphomolybdenum blue
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® ECO Phosphate, refill pack, test no. 5-84; Measuring range on PF-12Plus. Range on other photometers can be different.
- Dimensioner:170×115×70 mm (Including packaging)
- Til enhed:PF-12Plus;PF-3 Soil;Advance;PF-3 Fish;Visual
- Test pr. sæt:80
- Område:0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 • 5 mg/L (PO₄ - P); 0,2 - 5,0 mg/L (PO₄ - P)
- Indhold:Reagents for 80 tests in a box
- Cat. No.:MANA931284
- Platform:Colorimetric test kits
- Holdbarhed (måneder):36
- Vægt:129 g (Including packaging)
- Opbevaringstemperatur:15...25 °C
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



