Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Tests, DEHA

VISOCOLOR® Colorimetric Tests, DEHA

VISOCOLOR® ECO DEHA, refill pack, Reagents for 125 tests in a box, 0 • 0,01 • 0,03 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 mg/L (DEHA); 0,005 - 0,5 mg/L (DEHA); 0,01 - 0,60 mg/L (Carbo), Redox reaction, PF-12Plus; Advance; Visual, 170×115×70 mm (Including packaging), 125, 1 KIT
Katalog # MANA931224
Leverandør:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Tests, DEHA
Katalog # MANA931224
Leverandør:  MACHEREY-NAGEL
CAS nummer:  
VISOCOLOR® Colorimetric Tests, DEHA
VISOCOLOR® Colorimetric Tests, DEHA

VISOCOLOR® ECO DEHA, refill pack, Reagents for 125 tests in a box, 0 • 0,01 • 0,03 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 mg/L (DEHA); 0,005 - 0,5 mg/L (DEHA); 0,01 - 0,60 mg/L (Carbo), Redox reaction, PF-12Plus; Advance; Visual, 170×115×70 mm (Including packaging), 125, 1 KIT

CAS nummer:  
Price
645,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Reagents for the determination of DEHA in water samples.

  • Combines simple implementation with high measurement reliability

Simple determination directly on site or in the laboratory.

Sea water analysis: Yes

Hazardous material: No

Specifikationer

Specifikationer

  • Kemisk metode:
    Redox reaction
  • Pakn:
    1 KIT
  • Beskrivelse:
    VISOCOLOR® ECO DEHA, refill pack
  • Dimensioner:
    170×115×70 mm (Including packaging)
  • Til enhed:
    PF-12Plus; Advance; Visual
  • Test pr. sæt:
    125
  • Område:
    0 • 0,01 • 0,03 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 mg/L (DEHA); 0,005 - 0,5 mg/L (DEHA); 0,01 - 0,60 mg/L (Carbo)
  • Indhold:
    Reagents for 125 tests in a box
  • Cat. No.:
    MANA931224
  • Platform:
    Colorimetric test kits
  • Holdbarhed (måneder):
    12
  • Vægt:
    131,1 g (Including packaging)
  • Model:
    MANA931224
  • Opbevaringstemperatur:
    15...25 °C

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.