VISOCOLOR® Colorimetric Test, Ammonia
VISOCOLOR® ECO Ammonium 3, refill pack, test no. 5-08; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 50 tests in a box, 0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 mg/L (NH₄⁺); 0,1 - 2,5 mg/L (NH₄⁺), Indophenol, PF-12Plus; PF-3 Soil; Advance; PF-3 Fish; Visual, 170×115×70 mm (Including packaging), Sea water compatible, 50, 1 KIT
Leverandør: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Ammonia
Katalog # MANA931208
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Reagents for the determination of ammonia in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Hazardous material: Yes
Documents
Certifikater
Søg efter partinummerIFU VISOCOLOR® Alpha Ammonium NH₄⁺.pdf IFU VISOCOLOR® ECO Ammonium 15.pdf IFU VISOCOLOR® ECO Ammonium 3.pdf IFU VISOCOLOR® HE Ammonium.pdf IFU_Ammonium 15 ECO Visocolor
More
Specifikationer
Specifikationer
- Kemisk metode:Indophenol
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® ECO Ammonium 3, refill pack, test no. 5-08; Measuring range on PF-12Plus. Range on other photometers can be different
- Dimensioner:170×115×70 mm (Including packaging)
- Model:Sea water compatible
- Til enhed:PF-12Plus; PF-3 Soil; Advance; PF-3 Fish; Visual
- Test pr. sæt:50
- Område:0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 mg/L (NH₄⁺); 0,1 - 2,5 mg/L (NH₄⁺)
- Indhold:Reagents for 50 tests in a box
- Cat. No.:MANA931208
- Platform:Colorimetric test kits
- Holdbarhed (måneder):18
- Vægt:132 g (Including packaging)
- Model:MANA931208
- Opbevaringstemperatur:15...25 °C
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



