VISOCOLOR® Colorimetric Test, Phosphate
VISOCOLOR® ECO Phosphate, test no. 5-84; Measuring range on PF-12Plus. Range on other photometers can be different., Reagents for 80 tests, color chart and accessories in a box, 0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 • 5 mg/L (PO₄ - P); 0,2 - 5,0 mg/L (PO₄ - P), Phosphomolybdenum blue, PF-12Plus;PF-3 Soil;Advance;PF-3 Fish;Visual, 170×125×48 mm (Including packaging), 80, 1 KIT
Leverandør: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Phosphate
Katalog # MANA931084
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Reagents for the determination of phosphate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Specifikationer
Specifikationer
- Kemisk metode:Phosphomolybdenum blue
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® ECO Phosphate, test no. 5-84; Measuring range on PF-12Plus. Range on other photometers can be different.
- Dimensioner:170×125×48 mm (Including packaging)
- Til enhed:PF-12Plus;PF-3 Soil;Advance;PF-3 Fish;Visual
- Test pr. sæt:80
- Område:0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 • 5 mg/L (PO₄ - P); 0,2 - 5,0 mg/L (PO₄ - P)
- Indhold:Reagents for 80 tests, color chart and accessories in a box
- Cat. No.:MANA931084
- Platform:Colorimetric test kits
- Holdbarhed (måneder):36
- Vægt:212,85 g (Including packaging)
- Opbevaringstemperatur:15...25 °C



