VISOCOLOR® Colorimetric Test, Hardness
VISOCOLOR® ECO Carbonate hardness, Reagents for 100 tests and accessories in a box, 1 drop corresponds to 1 °e, Mixed indicator, Visual, 170×125×48 mm (Including packaging), 100, 1 KIT
Leverandør: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Hardness
Katalog # MANA931014
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Reagents for the determination of hardness in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
The term carbonate hardness describes the amount of magnesium and calcium ions for which equivalent molar amounts of hydrogen carbonate are available in the solution.
Hazardous material: Yes
Specifikationer
Specifikationer
- Kemisk metode:Mixed indicator
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® ECO Carbonate hardness
- Dimensioner:170×125×48 mm (Including packaging)
- Til enhed:Visual
- Test pr. sæt:100
- Område:1 drop corresponds to 1 °e
- Indhold:Reagents for 100 tests and accessories in a box
- Cat. No.:MANA931014
- Platform:Titrimetric test kits
- Holdbarhed (måneder):24
- Vægt:200,9 g (Including packaging)
- Opbevaringstemperatur:15...25 °C