VISOCOLOR® Colorimetric Test, Ammonia
VISOCOLOR® ECO Ammonium 3, test no. 5-08; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 50 tests, colour chart and accessories in a box, 0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 mg/L (NH₄⁺); 0,1 - 2,5 mg/L (NH₄⁺), Indophenol, PF-12Plus; PF-3 Soil; Advance; PF-3 Fish; Visual, 170×125×45 mm (Including packaging), Sea water compatible, 50, 1 KIT
Leverandør: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Ammonia
Katalog # MANA931008
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Reagents for the determination of ammonia in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Hazardous material: Yes
Documents
Certifikater
Søg efter partinummerIFU VISOCOLOR® Alpha Ammonium NH₄⁺.pdf IFU VISOCOLOR® ECO Ammonium 15.pdf IFU VISOCOLOR® ECO Ammonium 3.pdf IFU VISOCOLOR® HE Ammonium.pdf IFU_Ammonium 15 ECO Visocolor
More
Specifikationer
Specifikationer
- Kemisk metode:Indophenol
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® ECO Ammonium 3, test no. 5-08; Measuring range on PF-12Plus. Range on other photometers can be different
- Dimensioner:170×125×45 mm (Including packaging)
- Model:Sea water compatible
- Til enhed:PF-12Plus; PF-3 Soil; Advance; PF-3 Fish; Visual
- Test pr. sæt:50
- Område:0 • 0,2 • 0,3 • 0,5 • 0,7 • 1 • 2 • 3 mg/L (NH₄⁺); 0,1 - 2,5 mg/L (NH₄⁺)
- Indhold:Reagents for 50 tests, colour chart and accessories in a box
- Cat. No.:MANA931008
- Platform:Colorimetric test kits
- Holdbarhed (måneder):18
- Vægt:227,49 g (Including packaging)
- Model:MANA931008
- Opbevaringstemperatur:15...25 °C



