VISOCOLOR® Colorimetric Test, Phosphate
VISOCOLOR® HE Phosphate, refill pack, Reagents for 300 tests in a box, 0,0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,3 • 0,4 • 0,6 • 0,8 • 1,0 mg/L (PO₄ - P), Phosphomolybdenum blue, Visual, 235×130×47 mm (Including packaging), 300, 1 KIT
Leverandør: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Phosphate
Katalog # MANA920182
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Reagents for the determination of phosphate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Specifikationer
Specifikationer
- Kemisk metode:Phosphomolybdenum blue
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® HE Phosphate, refill pack
- Dimensioner:235×130×47 mm (Including packaging)
- Til enhed:Visual
- Test pr. sæt:300
- Område:0,0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,3 • 0,4 • 0,6 • 0,8 • 1,0 mg/L (PO₄ - P)
- Indhold:Reagents for 300 tests in a box
- Cat. No.:MANA920182
- Platform:Colorimetric test kits
- Holdbarhed (måneder):24
- Vægt:363,54 g (Including packaging)
- Opbevaringstemperatur:15...25 °C



