VISOCOLOR® Colorimetric Test, Phosphate
VISOCOLOR® HE Phosphate (DEV), refill pack, Reagents for 100 tests in a box, 0,0 • 0.01 • 0.02 • 0.03 • 0.05 • 0.07 • 0.10 • 0.15 • 0.20 • 0.25 mg/L (PO₄ - P), Phosphomolybdenum blue, Visual, 230×130×45 mm (Including packaging), 100, 1 KIT
Leverandør: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Phosphate
Katalog # MANA920180
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Reagents for the determination of phosphate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Specifikationer
Specifikationer
- Kemisk metode:Phosphomolybdenum blue
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® HE Phosphate (DEV), refill pack
- Dimensioner:230×130×45 mm (Including packaging)
- Til enhed:Visual
- Test pr. sæt:100
- Område:0,0 • 0.01 • 0.02 • 0.03 • 0.05 • 0.07 • 0.10 • 0.15 • 0.20 • 0.25 mg/L (PO₄ - P)
- Indhold:Reagents for 100 tests in a box
- Cat. No.:MANA920180
- Platform:Colorimetric test kits
- Holdbarhed (måneder):24
- Vægt:227 g (Including packaging)
- Opbevaringstemperatur:15...25 °C
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



