Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VISOCOLOR® Colorimetric Test, pH

VISOCOLOR® HE pH 4 - 10, refill pack, Reagents for 500 tests in a box, 4,0 • 5,0 • 5,5 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0 • 10,0, Mixed indicator, Visual, 237×130×45 mm (including packaging), Hazardous material: Yes, 500, 1 KIT
Katalog # MANA920174
Leverandør:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, pH
Katalog # MANA920174
Leverandør:  MACHEREY-NAGEL
CAS nummer:  
VISOCOLOR® Colorimetric Test, pH
VISOCOLOR® Colorimetric Test, pH

VISOCOLOR® HE pH 4 - 10, refill pack, Reagents for 500 tests in a box, 4,0 • 5,0 • 5,5 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0 • 10,0, Mixed indicator, Visual, 237×130×45 mm (including packaging), Hazardous material: Yes, 500, 1 KIT

CAS nummer:  
Price
736,00 kr
Log ind for tilgængelighed
1 * 1 Each
Specifikationer

Specifikationer

  • Kemisk metode:
    Mixed indicator
  • Pakn:
    1 KIT
  • Beskrivelse:
    VISOCOLOR® HE pH 4 - 10, refill pack
  • Dimensioner:
    237×130×45 mm (including packaging)
  • Model:
    Hazardous material: Yes
  • Til enhed:
    Visual
  • Test pr. sæt:
    500
  • Område:
    4,0 • 5,0 • 5,5 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0 • 10,0
  • Indhold:
    Reagents for 500 tests in a box
  • Cat. No.:
    MANA920174
  • Platform:
    Colorimetric test kits
  • Holdbarhed (måneder):
    24
  • Vægt:
    142 g (including packaging)
  • Model:
    MANA920174
  • Opbevaringstemperatur:
    15…25 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.