VISOCOLOR® Colorimetric Test, pH
VISOCOLOR® HE pH 4 - 10, Reagents for 500 tests, color chart and accessories in a box, 4,0 • 5,0 • 5,5 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0 • 10,0, Mixed indicator, Visual, 239×135×47 mm (including packaging), Hazardous material: Yes, 500, 1 KIT
Reagents for the determination of pH in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
- Kemisk metode:Mixed indicator
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® HE pH 4 - 10
- Dimensioner:239×135×47 mm (including packaging)
- Model:Hazardous material: Yes
- Til enhed:Visual
- Test pr. sæt:500
- Område:4,0 • 5,0 • 5,5 • 6,0 • 6,5 • 7,0 • 7,5 • 8,0 • 8,5 • 9,0 • 10,0
- Indhold:Reagents for 500 tests, color chart and accessories in a box
- Cat. No.:MANA920074
- Platform:Colorimetric test kits
- Holdbarhed (måneder):24
- Vægt:277,22 g (including packaging)
- Model:MANA920074
- Opbevaringstemperatur:15…25 °C



