VISOCOLOR® Colorimetric Test, Hardness
VISOCOLOR® HE Carbonate hardness C 20, refill pack, Reagents for 200 tests in a box, 0,5 - 20,0 - °e - (1 syringe filling); 0,2 - 7,2 - mmol/L (H⁺) (1 syringe filling), Mixed indicator, Visual, 170×115×70 mm (Including packaging), 200, 1 KIT
Leverandør: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Hardness
Katalog # MANA915203
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Reagents for the determination of hardness in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
The term carbonate hardness describes the amount of magnesium and calcium ions for which equivalent molar amounts of hydrogen carbonate are available in the solution.
Hazardous material: Yes
Specifikationer
Specifikationer
- Kemisk metode:Mixed indicator
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® HE Carbonate hardness C 20, refill pack
- Dimensioner:170×115×70 mm (Including packaging)
- Til enhed:Visual
- Test pr. sæt:200
- Område:0,5 - 20,0 - °e - (1 syringe filling); 0,2 - 7,2 - mmol/L (H⁺) (1 syringe filling)
- Indhold:Reagents for 200 tests in a box
- Cat. No.:MANA915203
- Platform:Titrimetric test kits
- Holdbarhed (måneder):24
- Vægt:236 g (Including packaging)
- Opbevaringstemperatur:15...25 °C
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



