VISOCOLOR® Colorimetric Test, Hardness
VISOCOLOR® HE Calcium CA 20, refill pack, Reagents for 200 tests in a box, 0,05 - 2,00 °e (1 syringe filling); 0,01 - 0,36 mmol/L (Ca²⁺) (1 syringe filling), Complexometric titration, Visual, 145×116×45 mm (Including packaging), 200, 1 KIT
Leverandør: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Hardness
Katalog # MANA915202
Leverandør: MACHEREY-NAGEL
CAS nummer:
Gå til:
Product Details & Documents
Reagents for the determination of hardness in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
The term carbonate hardness describes the amount of magnesium and calcium ions for which equivalent molar amounts of hydrogen carbonate are available in the solution.
Hazardous material: Yes
Specifikationer
Specifikationer
- Kemisk metode:Complexometric titration
- Pakn:1 KIT
- Beskrivelse:VISOCOLOR® HE Calcium CA 20, refill pack
- Dimensioner:145×116×45 mm (Including packaging)
- Til enhed:Visual
- Test pr. sæt:200
- Område:0,05 - 2,00 °e (1 syringe filling); 0,01 - 0,36 mmol/L (Ca²⁺) (1 syringe filling)
- Indhold:Reagents for 200 tests in a box
- Cat. No.:MANA915202
- Platform:Titrimetric test kits
- Holdbarhed (måneder):18
- Vægt:197,7 g (Including packaging)
- Opbevaringstemperatur:15...25 °C