HPLC columns, biochemical analysis, NUCLEOGEL® RP
Forkolonne, RP, 3,00 mm, 5 mm
Katalog # MANA719542
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
Polymer based reversed phase columns for biochemical applications.
- Polystyrene resin cross-linked with divinylbenzene
- Reversed phase application under extreme pH (pH stability 1 to 13)
- Eluent in column acetonitrile/water
NUCLEOGEL® RP columns with particle size of 100 Å are applied for reversed phase separations of small molecules such as pharmaceuticals with basic properties, nucleosides and nucleotides up to 5000 daltons; columns with 300 Å for large biomolecules (proteins, peptides etc.).
The Valco type guard column cartridge is used for analytical Valco column with the guard column holder B (MANA719539).
Specifikationer
- Kolonne form:Forkolonne
- Ø indv.:3,00 mm
- Længde:5 mm
- Seq:07
- Fase:RP
- Cat. No.:MANA719542
- USP code:L21
- Maks. tryk:180 bar
- pH-område:1 - 13



