HPLC columns, biochemical analysis, NUCLEOGEL® RP
Analytisk, RP, 100 Å, 5 µm, 4,60 mm, 150 mm
Katalog # MANA719454
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
Polymer based reversed phase columns for biochemical applications.
- Polystyrene resin cross-linked with divinylbenzene
- Reversed phase application under extreme pH (pH stability 1 to 13)
- Eluent in column acetonitrile/water
NUCLEOGEL® RP columns with particle size of 100 Å are applied for reversed phase separations of small molecules such as pharmaceuticals with basic properties, nucleosides and nucleotides up to 5000 daltons; columns with 300 Å for large biomolecules (proteins, peptides etc.).
The Valco type guard column cartridge is used for analytical Valco column with the guard column holder B (MANA719539).
Specifikationer
- Kolonne form:Analytisk
- Ø indv.:4,60 mm
- Partikelstørrelse:5 µm
- Porestørrelse:100 Å
- Længde:150 mm
- Seq:01
- Fase:RP
- Cat. No.:MANA719454
- USP code:L21
- Maks. tryk:180 bar
- pH-område:1 - 13
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



