Orders LinkContactUsLinkComponent
Glass wool
Glass wool
Quartz wool, very fine fibers
Katalog # MANA718587
Leverandør:   MACHEREY-NAGEL
Glass wool
Katalog # MANA718587
Leverandør:  MACHEREY-NAGEL
Glass wool
Glass wool
Quartz wool, very fine fibers
Price
643,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
This glass wool can be used for GC liners, packed GC columns etc.
Specifikationer
  • Beskrivelse:
    Quartz wool, very fine fibers
  • Seq:
    03