Orders LinkContactUsLinkComponent
Glass wool
Glass wool
Glass wool, long fibers, DMCS treated, for packed GC columns
Katalog # MANA706201
Leverandør:   MACHEREY-NAGEL
Glass wool
Katalog # MANA706201
Leverandør:  MACHEREY-NAGEL
Glass wool
Glass wool
Glass wool, long fibers, DMCS treated, for packed GC columns
Price
1.020,00 kr
Log in for availability
1 * 50 g
Product Details & Documents
This glass wool can be used for GC liners, packed GC columns etc.
Specifications
  • Beskrivelse:
    Glass wool, long fibers, DMCS treated, for packed GC columns
  • Seq:
    01