Extraction thimbles, borosilicate glass
27×55 mm, MN 649 R with collar, for dust analysis in industrial gases, exhaust gases and room air
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
Made from glass micro fibers without binder.
- Pollution monitoring, air sampling
- High temperature filtration (up to 500 °C)
- For solvents incompatible with cellulose thimbles (for example strong acids or bases)
Extraction thimbles for high temperatures and aggressive solvents
Specifikationer
- Indv. Ø×H:27×55 mm
- Model:MN 649 R with collar, for dust analysis in industrial gases, exhaust gases and room air