Orders LinkContactUsLinkComponent
Glass wool
Ashless, quantitative filter flocs, MN 2101
Katalog # MANA281120
Leverandør:   MACHEREY-NAGEL
Glass wool
Katalog # MANA281120
Leverandør:  MACHEREY-NAGEL
Glass wool
Glass wool
Ashless, quantitative filter flocs, MN 2101
Price
kr629.00
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Filter aids transform difficult precipitates and colloidal particles into a form which can be easily filtered. When slimy and strongly lyophilic, swelling precipitates are involved, the fibres of the filter flocs prevent formation of a continuous, impermeable layer on the filter. The resulting filter cake remains porous and permeable, and clogging of the filter is prevented.
Specifikationer
  • Beskrivelse:
    Ashless, quantitative filter flocs, MN 2101
  • Seq:
    03