Orders LinkContactUsLinkComponent
Tricine
Glass bottle for solids, 250 g
Katalog # A1085.0250
Leverandør:   PanReac AppliChem
Tricine
Katalog # A1085.0250
Leverandør:  PanReac AppliChem
CAS nummer:  5704-04-1
Tricine
Glass bottle for solids, 250 g
CAS nummer:  5704-04-1
Price
1.640,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
  • CAS nummer:5704-04-1
  • Formel:(HOCH₂)₃CNHCH₂CO₂H
  • MDL nummer:MFCD00004277
  • Molecular Weight:179.17 g/mol
  • Merck Index:13,09724
  • Smeltende Pt:187 °C
  • Tæthed:1,523 g/cm³ (19.85 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:227-193-6
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g
Specifikationer