Orders LinkContactUsLinkComponent
Tris-Borate-EDTA (TBE) buffer, 10X
Tris-Borate-EDTA (TBE) buffer, 10X
Tris-Borate-EDTA (TBE) buffer, 10X, 5 L
Katalog # A0972.5000PE
Leverandør:   PanReac AppliChem
Tris-Borate-EDTA (TBE) buffer, 10X
Katalog # A0972.5000PE
Leverandør:  PanReac AppliChem
CAS nummer:  
Tris-Borate-EDTA (TBE) buffer, 10X
Tris-Borate-EDTA (TBE) buffer, 10X, 5 L
CAS nummer:  
Price
993,00 kr
Log ind for tilgængelighed
1 * 5 L
Product Details & Documents
10X concentrated solution of 0,89 M Tris, 0,89 M boric acid and 0,02 M EDTA.Na₂, pH 8,3 ±0,2.
CT = Cubitainer®
Documents
Specifikationer
Specifikationer
  • Pakn:
    5 L
  • Beskrivelse:
    Tris-Borate-EDTA (TBE) buffer, 10X