Tris-Borate-EDTA (TBE) buffer, 10X
Tris-Borate-EDTA (TBE) buffer, 10X, 5 L
Katalog # A0972.5000PE
Leverandør: PanReac AppliChem
Gå til:
Product Details & Documents
10X concentrated solution of 0,89 M Tris, 0,89 M boric acid and 0,02 M EDTA.Na₂, pH 8,3 ±0,2.
CT = Cubitainer®
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:5 L
- Beskrivelse:Tris-Borate-EDTA (TBE) buffer, 10X