Rifampicin
Glass bottle for solids, 5 g
Katalog # A2220.0005
Leverandør: PanReac AppliChem
Gå til:
Product Details & Documents
Synonymer: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
- CAS nummer:13292-46-1
- Formel:C₄₃H₅₈N₄O₁₂
- MDL nummer:MFCD00151389
- Molecular Weight:822.95 g/mol
- Merck Index:12,08382
- Smeltende Pt:183…188 °C
- EINECS:236-312-0
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g



