Ribonuclease A
Glass bottle for solids, 1 g
Leverandør: PanReac AppliChem
Gå til:
Product Details & Documents
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g
- Cat. No.:A2760.1000
- CAS-nr:9001-99-4
- MDL No.:MFCD00132184
- Opbevaring betingelser:Freezer
- Enzym navn:Ribonuclease A
- Enzym synonymer:Rnase A
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



