Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Polymyxin Bsulfat

Polymyxin Bsulfat

Glass bottle for solids, 10 g
Katalog # A0890.0010
Leverandør:   PanReac AppliChem
Polymyxin Bsulfat
Katalog # A0890.0010
Leverandør:  PanReac AppliChem
CAS nummer:  1405-20-5
Polymyxin Bsulfat

Glass bottle for solids, 10 g

CAS nummer:  1405-20-5
Price
6.620,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
  • CAS nummer:1405-20-5
  • Formel:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
  • MDL nummer:MFCD00131991
  • Molecular Weight:1385.61 g/mol

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer