Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Polymyxin Bsulfat

Polymyxin Bsulfat

Glass bottle for solids, 1 g
Katalog # A0890.0001
Leverandør:   PanReac AppliChem
Polymyxin Bsulfat
Katalog # A0890.0001
Leverandør:  PanReac AppliChem
CAS nummer:  1405-20-5
Polymyxin Bsulfat

Glass bottle for solids, 1 g

CAS nummer:  1405-20-5
Price
641,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
  • CAS nummer:1405-20-5
  • Formel:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
  • MDL nummer:MFCD00131991
  • Molecular Weight:1385.61 g/mol

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.