Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES monohydrat

MES monohydrat

Glass bottle for solids, 1 kg
Catalog # A1074.1000
Supplier:   PanReac AppliChem
MES monohydrat
Catalog # A1074.1000
Supplier:  PanReac AppliChem
CAS Number:  145224-94-8
MES monohydrat

Glass bottle for solids, 1 kg

CAS Number:  145224-94-8
Price
5.410,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  • CAS Number:145224-94-8
  • Formula:C₆H₁₃NO₄S·1H₂O
  • MDL Number:MFCD00149409
  • Molecular Weight:213.25 g/mol
  • Merck Index:13,05929
  • Melting Pt:> 300 °C
  • Storage Temperature:Stue temperatur
  • EINECS:224-632-3
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifications