Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES

MES

Glass bottle for solids, 1 kg
Katalog # A0689.1000
Leverandør:   PanReac AppliChem
MES
Katalog # A0689.1000
Leverandør:  PanReac AppliChem
CAS nummer:  4432-31-9
MES

Glass bottle for solids, 1 kg

CAS nummer:  4432-31-9
Price
5.770,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid
  • CAS nummer:4432-31-9
  • Formel:C₆H₁₃NO₄S
  • MDL nummer:MFCD00006181
  • Molecular Weight:195.24 g/mol
  • Merck Index:13,05929
  • Smeltende Pt:316 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:224-632-3

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.