Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HEPES ≥99.5% suitable for buffer solutions

HEPES ≥99.5% suitable for buffer solutions

Plastic container, 10 kg
Katalog # A1069.9010
HEPES ≥99.5% suitable for buffer solutions
Katalog # A1069.9010
Leverandør:  
CAS nummer:  7365-45-9
HEPES ≥99.5% suitable for buffer solutions
HEPES ≥99.5% suitable for buffer solutions

Plastic container, 10 kg

CAS nummer:  7365-45-9
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
HEPES BUFFER GRADE 1 * 1 kg Katalog #A1069.1000

Produktet A1069.1000 ligner A1069.9010, men producenten og specifikationerne kan variere.

HEPES BUFFERQUALITY 1 * 25 kg Katalog #A1069.9025BW

Produktet A1069.9025BW ligner A1069.9010, men producenten og specifikationerne kan variere.

HEPES BUFFER GRADE 1 * 5 kg Katalog #A1069.5000

Produktet A1069.5000 ligner A1069.9010, men producenten og specifikationerne kan variere.

Se flere alternativer
Product Details & Documents
Synonymer:  HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
  • CAS nummer:7365-45-9
  • Formel:C₈H₁₈N₂O₄S
  • MDL nummer:MFCD00006158
  • Molecular Weight:238.31 g/mol
  • Merck Index:13,04672
  • Smeltende Pt:210…215 °C
  • Tæthed:1,448 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:230-907-9
Specifikationer

Specifikationstestresultater

  • Assay (titr.):
    min. 99.5 %
  • pH (1 %; H2O):
    4.7 - 6.0
  • Heavy metals (as Pb):
    max. 0.001 %
  • Water:
    max. 0.5 %
  • Chloride:
    max. 0.05 %
  • Sulfate:
    max. 0.05 %

Specifikationer

  • Pakning:
    Plastic container
  • Pakn:
    10 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.