Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

HEPES ≥99.5% suitable for buffer solutions

Plastic bottle for solids, 500 g
Catalog # A1069.0500
Supplier:   PanReac AppliChem
HEPES ≥99.5% suitable for buffer solutions
Catalog # A1069.0500
Supplier:  PanReac AppliChem
CAS Number:  7365-45-9
HEPES ≥99.5% suitable for buffer solutions
HEPES ≥99.5% suitable for buffer solutions

Plastic bottle for solids, 500 g

CAS Number:  7365-45-9
Price
1.940,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
  • CAS Number:7365-45-9
  • Formula:C₈H₁₈N₂O₄S
  • MDL Number:MFCD00006158
  • Molecular Weight:238.31 g/mol
  • Merck Index:13,04672
  • Melting Pt:210…215 °C
  • Density:1,448 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:230-907-9
Specifications

Specification Test Results

  • Assay (titr.):
    min. 99.5 %
  • pH (1 %; H2O):
    4.7 - 6.0
  • Heavy metals (as Pb):
    max. 0.001 %
  • Water:
    max. 0.5 %
  • Chloride:
    max. 0.05 %
  • Sulfate:
    max. 0.05 %

Specifications

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    500 g

Specifications