Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

HEPES ≥99.5% suitable for buffer solutions

Plastic bottle for solids, 100 g
  A1069.0100
HEPES ≥99.5% suitable for buffer solutions
  A1069.0100
 :  7365-45-9
HEPES ≥99.5% suitable for buffer solutions
HEPES ≥99.5% suitable for buffer solutions

Plastic bottle for solids, 100 g

 :  7365-45-9
 
507,00 kr
   
1 * 100 g
 
 :  HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
  •  :7365-45-9
  •  :C₈H₁₈N₂O₄S
  •  :MFCD00006158
  •  :238.31 g/mol
  •  :13,04672
  •  :210…215 °C
  •  :1,448 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :230-907-9
 

 

  • Assay (titr.):
    min. 99.5 %
  • pH (1 %; H2O):
    4.7 - 6.0
  • Heavy metals (as Pb):
    max. 0.001 %
  • Water:
    max. 0.5 %
  • Chloride:
    max. 0.05 %
  • Sulfate:
    max. 0.05 %

 

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    100 g

 

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.