Glycin
5 kg
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:13,04504
- Boiling Pt:181 °C (1013 hPa)
- Melting Pt:83 °C
- Density:1,64 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-272-2
Certificates
Search by Lot NumberSpecifications
Specifications
- Pakn:5 kg



