Orders LinkContactUsLinkComponent
Magnetic stirring bars, plain, PTFE
50 mm, 8 mm
Katalog # COWI001.250.7
Leverandør:   COWIE
Magnetic stirring bars, plain, PTFE
Katalog # COWI001.250.7
Leverandør:  COWIE
Magnetic stirring bars, plain, PTFE
Magnetic stirring bars, plain, PTFE
50 mm, 8 mm
Price
36,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.
Specifikationer
  • Ø:
    8 mm
  • Længde:
    50 mm
  • Seq:
    08