Orders LinkContactUsLinkComponent
Stoppers, flat type, PTFE
10/30, 40 mm
Katalog # COWI010.110
Leverandør:   COWIE
Stoppers, flat type, PTFE
Katalog # COWI010.110
Leverandør:  COWIE
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
10/30, 40 mm
Price
115,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure
Specifikationer
  • NS:
    10/30
  • Højde:
    40 mm