Orders LinkContactUsLinkComponent
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
14/35, 46 mm
Catalog # COWI010.114
Supplier:   COWIE
Stoppers, flat type, PTFE
Catalog # COWI010.114
Supplier:  COWIE
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
14/35, 46 mm
Price
160,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure
Specifications
  • NS:
    14/35
  • Højde:
    46 mm