Orders LinkContactUsLinkComponent
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
34 45, 60 mm
Katalog # COWI010.134
Leverandør:   COWIE
Stoppers, flat type, PTFE
Katalog # COWI010.134
Leverandør:  COWIE
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
34 45, 60 mm
Price
285,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure
Specifikationer
  • NS:
    34 45
  • Højde:
    60 mm