Capillaries for LC systems
Flexible capillaries, green, stainless steel, without fittings, 15,9 μl, 700 mm, I.Ø 0,17 mm
HEWL5065-9932
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Flexible capillaries, green, stainless steel, without fittings, 15,9 μl, 700 mm, I.Ø 0,17 mm
- Seq:99