Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tilbehør til pH-elektroder

Tilbehør til pH-elektroder

Sensor cable VP 6, max. Ø 7,5 mm, length 1 m
Katalog # 662-2081
Leverandør:   HAMILTON BONADUZ
Tilbehør til pH-elektroder
Katalog # 662-2081
Leverandør:  HAMILTON BONADUZ
Tilbehør til pH-elektroder

Sensor cable VP 6, max. Ø 7,5 mm, length 1 m

Price
1.860,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Sensor cable VP 6, max. Ø 7,5 mm, length 1 m
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.