Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sample vials, trident vials, squat or tall

Sample vials, trident vials, squat or tall

Squat, 21,25 ml, 25×66 mm, Yes
Katalog # 212-7069
Leverandør:   MURRAY S
Sample vials, trident vials, squat or tall
Katalog # 212-7069
Leverandør:  MURRAY S
Sample vials, trident vials, squat or tall
Sample vials, trident vials, squat or tall

Squat, 21,25 ml, 25×66 mm, Yes

Price
1.520,00 kr
Log ind for tilgængelighed
1 * 162 Items
Product Details & Documents

Vials made from neutral glass with PE plug stoppers. Suitable for a wide variety of uses in the laboratory, production, more.

  • Robust and economical, will withstand moderate temperature change
  • Wide opening for easy filling and emptying
  • Plug stopper for secure closure

Emballering: For production related reasons, the number of vials per box may vary.

Specifikationer
  • Kasket inkluderet:
    Yes
  • Diameter×H:
    25×66 mm
  • Kapacitet:
    21,25 ml
  • Beskrivelse:
    Squat