Jars with screw cap
with polypropylene screw cap, irradiated, individually wrapped, 150 ml
Leverandør: Anicrin
Gå til:
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
- Twist cap for quick opening and closing
- Very good temperature resistance
- Very good chemical resistance
Caution: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifikationer
- Model:with polypropylene screw cap, irradiated, individually wrapped
- Kapacitet:150 ml
- Seq:06