Orders LinkContactUsLinkComponent
Periodic Tables
Periodic Tables
Periodic table, multi-colour printed, German, 285×215 mm
Katalog # ROTH5412.1
Marketsource
Leverandør:   Roth Carl
Periodic Tables
Katalog # ROTH5412.1
Leverandør:  Roth Carl
Marketsource
Periodic Tables
Periodic table, multi-colour printed, German, 285×215 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These periodic tables of elements according to Fluck and Heumann allows for a clear presentation both in German or in English.
  • Multi-colour on both sides
  • Dirt proof shrink-wrapping
  • All relevant chemical and physical properties
Specifikationer
  • Beskrivelse:
    Periodic table, multi-colour printed, German
  • Seq:
    01
  • B×H:
    285×215 mm