X-Gal
Glass bottle for solids, 500 mg
Katalog # ROTH2315.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
- CAS nummer:7240-90-6
- Formel:C₁₄H₁₅BrClNO₆
- MDL nummer:MFCD00005666
- Molecular Weight:408.63 g/mol
- Smeltende Pt:237…239 °C
- Opbevaringstemperatur:Fryser
- EINECS:230-640-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



